missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-alpha-phenylglycinol, 99%

Catalog No. AAH2716306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2716306 5 g
AAH2716303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2716306 Supplier Thermo Scientific Chemicals Supplier No. H2716306

CAS: 102089-74-7 | C13H19NO3 | 237.299 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102089-74-7
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.299
MDL Number MFCD00274205
InChI Key IBDIOGYTZBKRGI-NSHDSACASA-N
Synonym boc-d-phenylglycinol, r-n-tert-butoxycarbonyl-2-phenylglycinol, --n-boc-d-alpha-phenylglycinol, r-tert-butyl 2-hydroxy-1-phenylethyl carbamate, n-boc-d-2-phenylglycinol, r-2-tert-butoxycarbonylamino-2-phenylethanol, tert-butyl 1r-2-hydroxy-1-phenylethyl carbamate, tert-butyl n-1r-2-hydroxy-1-phenylethyl carbamate, carbamic acid, 1r-2-hydroxy-1-phenylethyl-, 1,1-dimethylethyl ester, r---2-boc-amino-2-phenylethanol
PubChem CID 7016461
IUPAC Name tert-butyl N-[(1R)-2-hydroxy-1-phenylethyl]carbamate
SMILES CC(C)(C)OC(=O)NC(CO)C1=CC=CC=C1

Specifications

Melting Point 137°C to 141°C
Quantity 5 g
Optical Rotation −40° (c=1 in Chloroform)
Formula Weight 237.3
Percent Purity 99%
Chemical Name or Material N-Boc-D-alpha-phenylglycinol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only