Learn More
N-BOC-cis-4-Hydroxy-L-proline, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 87691-27-8 |
|---|---|
| Molecular Formula | C10H17NO5 |
| Molecular Weight (g/mol) | 231.25 |
| MDL Number | MFCD02094406 |
| InChI Key | BENKAPCDIOILGV-UHFFFAOYNA-N |
| Synonym | n-boc-cis-4-hydroxy-l-proline, 2s,4s-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, cis-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, 2s,4s-cis-1-n-boc-4-hydroxy-proline, boc-cishyp-oh, boc-cis-4-hydroxy-l-proline, cis-n-boc-4-hydroxy-l-proline, unii-h4ycj0kw0u, h4ycj0kw0u, 4s-1-tert-butoxycarbonyl-4-hydroxy-l-proline |
| PubChem CID | 688614 |
| IUPAC Name | 1-[(tert-butoxy)carbonyl]-4-hydroxypyrrolidine-2-carboxylic acid |
| SMILES | CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC441880010
|
N/A | 1g |
N/A
|
N/A | |||||
|
AC441880050
|
N/A | 5g |
N/A
|
N/A | |||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 87691-27-8 | |
| 231.25 | |
| BENKAPCDIOILGV-UHFFFAOYNA-N | |
| 688614 | |
| CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O |
| C10H17NO5 | |
| MFCD02094406 | |
| n-boc-cis-4-hydroxy-l-proline, 2s,4s-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, cis-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, 2s,4s-cis-1-n-boc-4-hydroxy-proline, boc-cishyp-oh, boc-cis-4-hydroxy-l-proline, cis-n-boc-4-hydroxy-l-proline, unii-h4ycj0kw0u, h4ycj0kw0u, 4s-1-tert-butoxycarbonyl-4-hydroxy-l-proline | |
| 1-[(tert-butoxy)carbonyl]-4-hydroxypyrrolidine-2-carboxylic acid |
Specifications
| 87691-27-8 | |
| 100.0 | |
| 97% | |
| C10H17NO5 | |
| − 49.60 | |
| BENKAPCDIOILGV-UHFFFAOYNA-N | |
| 1-[(tert-butoxy)carbonyl]-4-hydroxypyrrolidine-2-carboxylic acid | |
| 688614 | |
| 97% |
| 96.0 | |
| Authentic | |
| Glass Bottle | |
| MFCD02094406 | |
| n-boc-cis-4-hydroxy-l-proline, 2s,4s-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, cis-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, 2s,4s-cis-1-n-boc-4-hydroxy-proline, boc-cishyp-oh, boc-cis-4-hydroxy-l-proline, cis-n-boc-4-hydroxy-l-proline, unii-h4ycj0kw0u, h4ycj0kw0u, 4s-1-tert-butoxycarbonyl-4-hydroxy-l-proline | |
| CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O | |
| 231.25 | |
| 231.25 | |
| N-BOC-cis-4-Hydroxy-L-proline |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO – Research Use Only