missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-cis-4-hydroxy-L-proline, 97%

Catalog No. AAH2785003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2785003 1 g
AAH2785006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2785003 Supplier Thermo Scientific Chemicals Supplier No. H2785003

CAS: 87691-27-8 | C10H17NO5 | 231.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87691-27-8
Molecular Formula C10H17NO5
Molecular Weight (g/mol) 231.25
MDL Number MFCD02094406
InChI Key BENKAPCDIOILGV-UHFFFAOYNA-N
Synonym n-boc-cis-4-hydroxy-l-proline, 2s,4s-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, cis-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, 2s,4s-cis-1-n-boc-4-hydroxy-proline, boc-cishyp-oh, boc-cis-4-hydroxy-l-proline, cis-n-boc-4-hydroxy-l-proline, unii-h4ycj0kw0u, h4ycj0kw0u, 4s-1-tert-butoxycarbonyl-4-hydroxy-l-proline
PubChem CID 688614
IUPAC Name (2S,4S)-4-hydroxy-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O

Specifications

Melting Point ∽146°C (decomposition)
Quantity 1 g
Formula Weight 231.25
Percent Purity 97%
Chemical Name or Material N-Boc-cis-4-hydroxy-L-proline
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 627-315-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only