missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-4-Piperidone, 99%

Catalog No. p-4258793
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC330271000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC330271000 Supplier Thermo Scientific Chemicals Supplier No. 330271000

CAS: 79099-07-3 | C10H17NO3 | 199.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79099-07-3
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD00151800
InChI Key ROUYFJUVMYHXFJ-UHFFFAOYSA-N
Synonym 1-boc-4-piperidone, n-boc-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, 1-boc-4-piperidinone, boc-4-piperidone, 1-tert-butoxycarbonyl-4-piperidone, n-boc-4-piperidinone, boc-piperidone, 1-t-boc-4-piperidone
PubChem CID 735900
IUPAC Name tert-butyl 4-oxopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(=O)CC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 74.0°C to 75.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: slightly soluble
Formula Weight 199.25
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-BOC-4-Piperidone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

RUO – Research Use Only