missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-4-Piperidone, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 330270250

Numéro de catalogue. AC330270250

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spécifications

Spécifications

N-BOC-4-Piperidone
98.5
Authentic
Glass bottle
MFCD00151800
1-boc-4-piperidone, n-boc-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, 1-boc-4-piperidinone, boc-4-piperidone, 1-tert-butoxycarbonyl-4-piperidone, n-boc-4-piperidinone, boc-piperidone, 1-t-boc-4-piperidone
ROUYFJUVMYHXFJ-UHFFFAOYSA-N
tert-butyl 4-oxopiperidine-1-carboxylate
735900
99%
79099-07-3
100.0
98.5% min. (GC)
C10H17NO3
25g
Solubility in water: slightly soluble
CC(C)(C)OC(=O)N1CCC(=O)CC1
199.25
199.25
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

FDS
Documents

Documents

Promotions

Promotions