missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-oxo-L-proline tert-butyl ester, 97%

Catalog No. p-7029862
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2737503 1 g
AAH2737506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2737503 Supplier Thermo Scientific Chemicals Supplier No. H2737503
Only null left

CAS: 166410-05-5 | C14H23NO5 | 285.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 166410-05-5
Molecular Formula C14H23NO5
Molecular Weight (g/mol) 285.34
MDL Number MFCD06797013
InChI Key MPNWQUWKRDADHK-JTQLQIEISA-N
Synonym n-boc-4-oxo-l-proline tert-butyl ester, s-di-tert-butyl 4-oxopyrrolidine-1,2-dicarboxylate, boc-4-oxo-l-proline tert-butyl ester, 1,2-di-tert-butyl 2s-4-oxopyrrolidine-1,2-dicarboxylate, boc-4-oxo-l-prolinetert-butylester, di-tert-butyl s-4-oxopyrrolidine-1,2-dicarboxylate, di-tert-butyl 2s-4-oxopyrrolidine-1,2-dicarboxylate, 1-tert-butoxycarbonyl-4-oxo-l-proline tert-butyl ester, 1-tert-butyloxycarbonyl-4-oxo-l-proline tert-butyl ester, s-4-oxopyrrolidine-1,2-dicarboxylic acid di-tert-butyl ester
PubChem CID 11346670
IUPAC Name ditert-butyl (2S)-4-oxopyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)C1CC(=O)CN1C(=O)OC(C)(C)C

Specifications

Melting Point 64°C to 67°C
Quantity 1 g
Formula Weight 285.35
Percent Purity 97%
Chemical Name or Material N-Boc-4-oxo-L-proline tert-butyl ester
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only