missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-methoxy-D-phenylalanine, 95%

Catalog No. p-7034398
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5208203 1 g
AAH5208206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5208203 Supplier Thermo Scientific Chemicals Supplier No. H5208203
Only null left

CAS: 68856-96-2 | C15H21NO5 | 295.335 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68856-96-2
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.335
InChI Key SLWWWZWJISHVOU-GFCCVEGCSA-N
Synonym boc-d-tyr me-oh, boc-4-methoxy-d-phenylalanine, boc-o-methyl-d-tyrosine, boc-4-methoxy-d-phe-oh, n-boc-4-methoxy-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl propanoic acid, d-tyrosine,n-1,1-dimethylethoxy carbonyl-o-methyl, d-tyrosine, n-1,1-dimethylethoxy carbonyl-o-methyl, boc-l-phe 4-och3-oh, boc-d-phe 4-ome-oh
PubChem CID 7017908
IUPAC Name (2R)-3-(4-methoxyphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)OC)C(=O)O

Specifications

Quantity 1 g
Formula Weight 295.33
Percent Purity 95%
Chemical Name or Material N-Boc-4-methoxy-D-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only