missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-iodo-L-phenylalanine, 98%

Catalog No. aah5196006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5196006 5 g
AAH5196014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5196006 Supplier Thermo Scientific Chemicals Supplier No. H5196006

CAS: 62129-44-6 | C14H18INO4 | 391.21 g/mol

N-Boc-4-iodo-L-phenylalanine is used as pharmaceutical intermediate. It is also used as a p-iodo phenyl alanine derivative with N-Boc protection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-4-iodo-L-phenylalanine is used as pharmaceutical intermediate. It is also used as a p-iodo phenyl alanine derivative with N-Boc protection.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 62129-44-6
Molecular Formula C14H18INO4
Molecular Weight (g/mol) 391.21
MDL Number MFCD00037119
InChI Key JZLZDBGQWRBTHN-NSHDSACASA-N
Synonym boc-4-iodo-l-phenylalanine, boc-phe 4-i-oh, boc-l-4-iodophenylalanine, boc-l-phe 4-i-oh, s-2-tert-butoxycarbonyl amino-3-4-iodophenyl propanoic acid, boc-4'-iodo-l-phe, 2s-2-tert-butoxy carbonyl amino-3-4-iodophenyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-4-iodophenyl propanoic acid, l-n-tert-butoxycarbonyl-4-iodophenylalanine, n-tert-butoxycarbonyl-4-iodo-l-phenylalanine
PubChem CID 2755956
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(I)C=C1)C(O)=O

Specifications

Melting Point 150°C (decomposition)
Quantity 5 g
Beilstein 4503851
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Optical Rotation +22.5° (c=1 in Ethyl Acetate)
IUPAC Name (2S)-3-(4-iodophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Formula Weight 391.21
Percent Purity 98%
Chemical Name or Material N-Boc-4-iodo-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only