missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-4-Hydroxypiperidine, 97%, Thermo Scientific™

Catalog No. p-2691897
Change view
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC368270050 5g Glass bottle
AC368270250 25g Glass bottle
AC368271000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC368270050 Supplier Thermo Scientific Chemicals Supplier No. 368270050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 109384-19-2
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.27
MDL Number MFCD01075174
InChI Key PWQLFIKTGRINFF-UHFFFAOYSA-N
Synonym n-boc-4-hydroxypiperidine, 1-boc-4-hydroxypiperidine, n-boc-4-piperidinol, 1-boc-4-piperidinol, n-boc-4-hydroxylpiperidine, n-boc-4-hydroxy piperidine, 1-tert-butoxycarbonyl-4-hydroxypiperidine, 4-hydroxy-n-boc-piperidine, 1-boc-4-hydroxy-piperidine
PubChem CID 643502
IUPAC Name tert-butyl 4-hydroxypiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5g
Formula Weight 201.27
Percent Purity 97%
Chemical Name or Material N-BOC-4-Hydroxypiperidine, 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only