missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-chloro-D-phenylalanine, 95%

CAS: 57292-44-1 | C14H18ClNO4 | 299.75 g/mol

Supplier:  Thermo Scientific Chemicals H5218106

 View more versions of this product

Catalog No. AAH5218106


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Boc-4-chloro-D-phenylalanine
110°C
MFCD00076978
5381988
BETBOAZCLSJOBQ-LLVKDONJSA-N
CC(C)(C)OC(=O)N[C@H](CC1=CC=C(Cl)C=C1)C(O)=O
299.75
299.75
57292-44-1
C14H18ClNO4
5 g
boc-d-phe 4-cl-oh, boc-4-chloro-d-phenylalanine, boc-d-4-chlorophe, boc-d-4-chlorophenylalanine, n-tert-butoxycarbonyl-4-chloro-d-phenylalanine, n-boc-4-chloro-d-phenylalanine, r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, r-n-boc-4-chlorophenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, d-phenylalanine, 4-chloro-n-1,1-dimethylethoxy carbonyl
−26° (c=1 in Ethyl Acetate)
(2R)-3-(4-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
7018791
95%

Chemical Identifiers

57292-44-1
299.75
BETBOAZCLSJOBQ-LLVKDONJSA-N
7018791
CC(C)(C)OC(=O)N[C@H](CC1=CC=C(Cl)C=C1)C(O)=O
C14H18ClNO4
MFCD00076978
boc-d-phe 4-cl-oh, boc-4-chloro-d-phenylalanine, boc-d-4-chlorophe, boc-d-4-chlorophenylalanine, n-tert-butoxycarbonyl-4-chloro-d-phenylalanine, n-boc-4-chloro-d-phenylalanine, r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, r-n-boc-4-chlorophenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, d-phenylalanine, 4-chloro-n-1,1-dimethylethoxy carbonyl
(2R)-3-(4-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only