missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-chloro-D-phenylalanine, 95%

Catalog No. AAH52181MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52181MD 250 mg
AAH5218103 1 g
AAH5218106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52181MD Supplier Thermo Scientific Chemicals Supplier No. H52181MD

CAS: 57292-44-1 | C14H18ClNO4 | 299.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57292-44-1
Molecular Formula C14H18ClNO4
Molecular Weight (g/mol) 299.75
MDL Number MFCD00076978
InChI Key BETBOAZCLSJOBQ-LLVKDONJSA-N
Synonym boc-d-phe 4-cl-oh, boc-4-chloro-d-phenylalanine, boc-d-4-chlorophe, boc-d-4-chlorophenylalanine, n-tert-butoxycarbonyl-4-chloro-d-phenylalanine, n-boc-4-chloro-d-phenylalanine, r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, r-n-boc-4-chlorophenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, d-phenylalanine, 4-chloro-n-1,1-dimethylethoxy carbonyl
PubChem CID 7018791
IUPAC Name (2R)-3-(4-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CC1=CC=C(Cl)C=C1)C(O)=O

Specifications

Melting Point 110°C
Quantity 250 mg
Beilstein 5381988
Optical Rotation −26° (c=1 in Ethyl Acetate)
Formula Weight 299.75
Percent Purity 95%
Chemical Name or Material N-Boc-4-chloro-D-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only