missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-bromo-L-phenylalanine, 98%

Catalog No. AAH5196906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5196906 5 g
AAH5196903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5196906 Supplier Thermo Scientific Chemicals Supplier No. H5196906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 62129-39-9 | C14H17BrNO4 | 343.20 g/mol

N-Boc-4-bromo-L-phenylalanine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-4-bromo-L-phenylalanine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 62129-39-9
Molecular Formula C14H17BrNO4
Molecular Weight (g/mol) 343.20
MDL Number MFCD00237571
InChI Key ULNOXUAEIPUJMK-NSHDSACASA-M
Synonym s-n-boc-4-bromophenylalanine, boc-l-4-bromophenylalanine, boc-phe 4-br-oh, boc-4-bromo-l-phenylalanine, boc-l-4-bromophe, boc-p-bromo-phe-oh, boc-4'-bromo-l-phe, n-boc-4-bromo-l-phenylalanine, s-3-4-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, boc-l-4-bromophenylalnine
PubChem CID 2734477
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(Br)C=C1)C([O-])=O

Specifications

Melting Point 118°C to 120°C
Quantity 5 g
Solubility Information Slightly soluble in water.
IUPAC Name (2S)-3-(4-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Formula Weight 344.2
Percent Purity 98%
Chemical Name or Material N-Boc-4-bromo-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only