missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4,4-difluoro-L-proline, 97+%, Thermo Scientific Chemicals

Catalog No. AAH2705203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2705203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2705203 Supplier Thermo Scientific Chemicals Supplier No. H2705203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 203866-15-3 | C10H15F2NO4 | 251.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 203866-15-3
Molecular Formula C10H15F2NO4
Molecular Weight (g/mol) 251.23
MDL Number MFCD03094917
InChI Key WTMZYKCXBXPVPT-UHFFFAOYNA-N
Synonym n-boc-4,4-difluoro-l-proline, s-1-tert-butoxycarbonyl-4,4-difluoropyrrolidine-2-carboxylic acid, n-t-boc-4,4-difluoro-l-proline, 1-tert-butoxycarbonyl-4,4-difluoro-l-proline, 2s-1-tert-butoxycarbonyl-4,4-difluoropyrrolidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl-4,4-difluoropyrrolidine-2-carboxylic acid, boc-4,4-difluoro-l-proline, s-1-boc-4,4-difluoropyrrolidine-2-carboxylic acid, n-boc-4,4-difluoroproline
PubChem CID 22977458
SMILES CC(C)(C)OC(=O)N1CC(F)(F)CC1C(O)=O

Specifications

Melting Point 120°C to 124°C
Quantity 1 g
IUPAC Name 1-[(tert-butoxy)carbonyl]-4,4-difluoropyrrolidine-2-carboxylic acid
Formula Weight 251.24
Percent Purity ≥97%
Chemical Name or Material N-Boc-4,4-difluoro-L-proline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only