missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-3-hydroxy-L-valine, 97%
CAS: 102507-13-1 | C10H19NO5 | 233.26 g/mol
$584.32 - $1737.05
Chemical Identifiers
| CAS | 102507-13-1 |
|---|---|
| Molecular Formula | C10H19NO5 |
| Molecular Weight (g/mol) | 233.26 |
| MDL Number | MFCD03094792 |
| InChI Key | SZVRVSZFEDIMFM-ZCFIWIBFSA-N |
| Synonym | s-2-tert-butoxycarbonyl amino-3-hydroxy-3-methylbutanoic acid, n-boc-s-2-amino-3-hydroxy-3-methylbutanoic acid, s-2-n-boc-amino-3-hydroxy-3-methylbutyric acid, s-n-boc-2-amino-3-hydroxy-3-methylbutanoic acid, n-tert-butoxycarbonyl-3-hydroxy-l-valine, 2s-2-n-tert-butoxycarbonyl amino-3-hydroxy-3-methylbutanoic acid, n-boc-2-amino-3-hydroxy-3-methylbutanoic acid, s-2-boc-amino-3-hydroxy-3-methylbutyric acid, boc-s-2-amino-3-hydroxy-3-methylbutanoic acid, l-valine, n-1,1-dimethylethoxy carbonyl-3-hydroxy |
| PubChem CID | 11075314 |
| IUPAC Name | (2S)-3-hydroxy-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid |
| SMILES | CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH52429MD
|
Thermo Scientific Chemicals
H52429MD |
250 mg |
Each for $584.32
|
|
|||||
|
AAH5242903
|
Thermo Scientific Chemicals
H5242903 |
1 g |
Each for $1,737.05
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 102507-13-1 | |
| 233.26 | |
| SZVRVSZFEDIMFM-ZCFIWIBFSA-N | |
| 11075314 | |
| CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)O |
| C10H19NO5 | |
| MFCD03094792 | |
| s-2-tert-butoxycarbonyl amino-3-hydroxy-3-methylbutanoic acid, n-boc-s-2-amino-3-hydroxy-3-methylbutanoic acid, s-2-n-boc-amino-3-hydroxy-3-methylbutyric acid, s-n-boc-2-amino-3-hydroxy-3-methylbutanoic acid, n-tert-butoxycarbonyl-3-hydroxy-l-valine, 2s-2-n-tert-butoxycarbonyl amino-3-hydroxy-3-methylbutanoic acid, n-boc-2-amino-3-hydroxy-3-methylbutanoic acid, s-2-boc-amino-3-hydroxy-3-methylbutyric acid, boc-s-2-amino-3-hydroxy-3-methylbutanoic acid, l-valine, n-1,1-dimethylethoxy carbonyl-3-hydroxy | |
| (2S)-3-hydroxy-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid |
Specifications
| 102507-13-1 | |
| MFCD03094792 | |
| s-2-tert-butoxycarbonyl amino-3-hydroxy-3-methylbutanoic acid, n-boc-s-2-amino-3-hydroxy-3-methylbutanoic acid, s-2-n-boc-amino-3-hydroxy-3-methylbutyric acid, s-n-boc-2-amino-3-hydroxy-3-methylbutanoic acid, n-tert-butoxycarbonyl-3-hydroxy-l-valine, 2s-2-n-tert-butoxycarbonyl amino-3-hydroxy-3-methylbutanoic acid, n-boc-2-amino-3-hydroxy-3-methylbutanoic acid, s-2-boc-amino-3-hydroxy-3-methylbutyric acid, boc-s-2-amino-3-hydroxy-3-methylbutanoic acid, l-valine, n-1,1-dimethylethoxy carbonyl-3-hydroxy | |
| CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)O | |
| 233.26 | |
| 233.26 | |
| N-Boc-3-hydroxy-L-valine |
| C10H19NO5 | |
| 250 mg | |
| SZVRVSZFEDIMFM-ZCFIWIBFSA-N | |
| (2S)-3-hydroxy-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid | |
| 11075314 | |
| 97% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only