missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-hydroxy-L-valine, 97%

Catalog No. AAH52429MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52429MD 250 mg
AAH5242903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52429MD Supplier Thermo Scientific Chemicals Supplier No. H52429MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 102507-13-1 | C10H19NO5 | 233.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102507-13-1
Molecular Formula C10H19NO5
Molecular Weight (g/mol) 233.26
MDL Number MFCD03094792
InChI Key SZVRVSZFEDIMFM-ZCFIWIBFSA-N
Synonym s-2-tert-butoxycarbonyl amino-3-hydroxy-3-methylbutanoic acid, n-boc-s-2-amino-3-hydroxy-3-methylbutanoic acid, s-2-n-boc-amino-3-hydroxy-3-methylbutyric acid, s-n-boc-2-amino-3-hydroxy-3-methylbutanoic acid, n-tert-butoxycarbonyl-3-hydroxy-l-valine, 2s-2-n-tert-butoxycarbonyl amino-3-hydroxy-3-methylbutanoic acid, n-boc-2-amino-3-hydroxy-3-methylbutanoic acid, s-2-boc-amino-3-hydroxy-3-methylbutyric acid, boc-s-2-amino-3-hydroxy-3-methylbutanoic acid, l-valine, n-1,1-dimethylethoxy carbonyl-3-hydroxy
PubChem CID 11075314
IUPAC Name (2S)-3-hydroxy-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)O

Specifications

Quantity 250 mg
Formula Weight 233.26
Percent Purity 97%
Chemical Name or Material N-Boc-3-hydroxy-L-valine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only