Learn More
N-Boc-3-dimethylamino-L-alanine, 97%
CAS: 94778-71-9 | C10H20N2O4 | 232.28 g/mol
Supplier: Thermo Scientific Chemicals H52810MD
Description
N-Boc-3-dimethylamino-L-alanine is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-3-dimethylamino-L-alanine is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
| N-Boc-3-dimethylamino-L-alanine | |
| C10H20N2O4 | |
| boc-aza-l-leucine, 2s-2-tert-butoxy carbonyl amino-3-dimethylamino propanoic acid, n-tert-butoxycarbonyl-3-dimethylamino-l-alanine, 2s-2-tert-butoxycarbonyl amino-3-dimethylamino propanoic acid, ?-n,n-dimethylamino-l-ala, ?-n,n-dimethylamino-l-ala, na-boc-s-2-amino-3-dimethylamino propionic acid, n-alpha-boc-s-2-amino-3-dimethylamino propionic acid, s-2-tert-butoxycarbonylamino-3-dimethylamino-propionic acid, l-alanine,3-dimethylamino-n-1,1-dimethylethoxy carbonyl, l-alanine,3-dimethylamino-n-1,1-dimethylethoxy carbonyl | |
| CC(C)(C)OC(=O)NC(CN(C)C)C(=O)O | |
| 250 mg | |
| 17998956 | |
| 97% |
| 94778-71-9 | |
| MFCD02682423 | |
| VCDQZVYJKDSORW-ZETCQYMHSA-N | |
| (2S)-3-(dimethylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
| 232.28 | |
| 232.28 |
Chemical Identifiers
| 94778-71-9 | |
| 232.28 | |
| VCDQZVYJKDSORW-ZETCQYMHSA-N | |
| 17998956 | |
| CC(C)(C)OC(=O)NC(CN(C)C)C(=O)O |
| C10H20N2O4 | |
| MFCD02682423 | |
| boc-aza-l-leucine, 2s-2-tert-butoxy carbonyl amino-3-dimethylamino propanoic acid, n-tert-butoxycarbonyl-3-dimethylamino-l-alanine, 2s-2-tert-butoxycarbonyl amino-3-dimethylamino propanoic acid, ?-n,n-dimethylamino-l-ala, ?-n,n-dimethylamino-l-ala, na-boc-s-2-amino-3-dimethylamino propionic acid, n-alpha-boc-s-2-amino-3-dimethylamino propionic acid, s-2-tert-butoxycarbonylamino-3-dimethylamino-propionic acid, l-alanine,3-dimethylamino-n-1,1-dimethylethoxy carbonyl, l-alanine,3-dimethylamino-n-1,1-dimethylethoxy carbonyl | |
| (2S)-3-(dimethylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only