missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-3-cyclohexyl-L-alanine, 98%
CAS: 37736-82-6 | C14H25NO4 | 271.357 g/mol
$144.59 - $483.70
Chemical Identifiers
| CAS | 37736-82-6 |
|---|---|
| Molecular Formula | C14H25NO4 |
| Molecular Weight (g/mol) | 271.357 |
| MDL Number | MFCD00211335 |
| InChI Key | MSZQAQJBXGTSHP-NSHDSACASA-N |
| Synonym | boc-cha-oh, boc-cha-oh hydrate, s-2-tert-butoxycarbonyl amino-3-cyclohexylpropanoic acid, boc-beta-cyclohexyl-ala-oh, boc-l-cyclohexylalanine, s-2-tert-butoxycarbonylamino-3-cyclohexyl-propionic acid, n-boc-b-cyclohexylalanine, 2s-2-tert-butoxycarbonyl amino-3-cyclohexylpropanoic acid, boc-cyclohexyl-ala-oh, boc-3-cyclohexyl-l-alanine |
| PubChem CID | 10588189 |
| IUPAC Name | (2S)-3-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
| SMILES | CC(C)(C)OC(=O)NC(CC1CCCCC1)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5528203
|
Thermo Scientific Chemicals
H5528203 |
1 g |
Each for $144.59
|
|
|||||
|
AAH5528206
|
Thermo Scientific Chemicals
H5528206 |
5 g |
Each for $483.70
|
|
|||||
Description
It is an agrochemical, organic and pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an agrochemical, organic and pharmaceutical intermediates.
Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
Chemical Identifiers
| 37736-82-6 | |
| 271.357 | |
| MSZQAQJBXGTSHP-NSHDSACASA-N | |
| 10588189 | |
| CC(C)(C)OC(=O)NC(CC1CCCCC1)C(=O)O |
| C14H25NO4 | |
| MFCD00211335 | |
| boc-cha-oh, boc-cha-oh hydrate, s-2-tert-butoxycarbonyl amino-3-cyclohexylpropanoic acid, boc-beta-cyclohexyl-ala-oh, boc-l-cyclohexylalanine, s-2-tert-butoxycarbonylamino-3-cyclohexyl-propionic acid, n-boc-b-cyclohexylalanine, 2s-2-tert-butoxycarbonyl amino-3-cyclohexylpropanoic acid, boc-cyclohexyl-ala-oh, boc-3-cyclohexyl-l-alanine | |
| (2S)-3-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
| 37736-82-6 | |
| MFCD00211335 | |
| 3060603 | |
| MSZQAQJBXGTSHP-NSHDSACASA-N | |
| (2S)-3-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
| 10588189 | |
| 98% |
| C14H25NO4 | |
| 1 g | |
| boc-cha-oh, boc-cha-oh hydrate, s-2-tert-butoxycarbonyl amino-3-cyclohexylpropanoic acid, boc-beta-cyclohexyl-ala-oh, boc-l-cyclohexylalanine, s-2-tert-butoxycarbonylamino-3-cyclohexyl-propionic acid, n-boc-b-cyclohexylalanine, 2s-2-tert-butoxycarbonyl amino-3-cyclohexylpropanoic acid, boc-cyclohexyl-ala-oh, boc-3-cyclohexyl-l-alanine | |
| CC(C)(C)OC(=O)NC(CC1CCCCC1)C(=O)O | |
| 271.357 | |
| 271.36 | |
| N-Boc-3-cyclohexyl-L-alanine |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only