missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-cyano-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog No. AAH6251703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6251703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6251703 Supplier Thermo Scientific Chemicals Supplier No. H6251703
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 131980-30-8
Molecular Formula C15H18N2O4
Molecular Weight (g/mol) 290.319
MDL Number MFCD00797560
InChI Key FDQDHMZKOPOWFE-LBPRGKRZSA-N
Synonym boc-phe 3-cn-oh, boc-l-3-cyanophenylalanine, boc-3-cyano-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid, boc-l-3-cn-phe-oh, 2s-2-tert-butoxy carbonylamino-3-3-cyanophenyl propanoic acid, boc-l-3-cyanophe, boc-l-3-cyano phenylalanine
PubChem CID 2734498
IUPAC Name (2S)-3-(3-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC(=C1)C#N)C(=O)O

Specifications

UN Number UN3439
Quantity 1 g
Formula Weight 290.31
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-3-cyano-L-phenylalanine
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only