missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-chloro-L-phenylalanine, 95%

Catalog No. p-7034334
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5201503 1 g
AAH5201506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5201503 Supplier Thermo Scientific Chemicals Supplier No. H5201503
Only null left

CAS: 114873-03-9 | C14H18ClNO4 | 299.751 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 114873-03-9
Molecular Formula C14H18ClNO4
Molecular Weight (g/mol) 299.751
MDL Number MFCD00672515
InChI Key RCZHBTHQISEPPP-NSHDSACASA-N
Synonym boc-3-chloro-l-phenylalanine, boc-phe 3-cl-oh, boc-l-3-chlorophenylalanine, s-n-boc-3-chlorophenylalanine, boc-l-phe 3-cl-oh, boc-l-3-chlorophe, boc-d-3-chlorophenylalanine, boc-l-3-chloro-phe-oh, s-2-tert-butoxycarbonyl amino-3-3-chlorophenyl propanoic acid, n-boc-3-chloro-l-phenylalanine
PubChem CID 2761465
IUPAC Name (2S)-3-(3-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)Cl)C(=O)O

Specifications

Quantity 1 g
Formula Weight 299.75
Percent Purity 95%
Chemical Name or Material N-Boc-3-chloro-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only