missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-3-bromo-L-phenylalanine, 95%
CAS: 82278-73-7 | C14H18BrNO4 | 344.205 g/mol
$215.37 - $714.95
Chemical Identifiers
| CAS | 82278-73-7 |
|---|---|
| Molecular Formula | C14H18BrNO4 |
| Molecular Weight (g/mol) | 344.205 |
| MDL Number | MFCD01317710 |
| InChI Key | FBUDYESOPLBQIR-NSHDSACASA-N |
| Synonym | s-n-boc-3-bromophenylalanine, boc-l-3-bromophenylalanine, boc-3-bromo-l-phenylalanine, boc-phe 3-br-oh, boc-l-3-br-phe-oh, boc-l-3-bromophe, n-boc-3-bromo-l-phenylalanine, s-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-2-tert-butoxy carbonylamino-3-3-bromophenyl propanoic acid |
| PubChem CID | 2734476 |
| IUPAC Name | (2S)-3-(3-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
| SMILES | CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)Br)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5202403
|
Thermo Scientific Chemicals
H5202403 |
1 g |
Each for $215.37
|
|
|||||
|
AAH5202406
|
Thermo Scientific Chemicals
H5202406 |
5 g |
Each for $714.95
|
|
|||||
Description
N-Boc-3-bromo-L-phenylalanine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-3-bromo-L-phenylalanine is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 82278-73-7 | |
| 344.205 | |
| FBUDYESOPLBQIR-NSHDSACASA-N | |
| 2734476 | |
| CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)Br)C(=O)O |
| C14H18BrNO4 | |
| MFCD01317710 | |
| s-n-boc-3-bromophenylalanine, boc-l-3-bromophenylalanine, boc-3-bromo-l-phenylalanine, boc-phe 3-br-oh, boc-l-3-br-phe-oh, boc-l-3-bromophe, n-boc-3-bromo-l-phenylalanine, s-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-2-tert-butoxy carbonylamino-3-3-bromophenyl propanoic acid | |
| (2S)-3-(3-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
| 82278-73-7 | |
| MFCD01317710 | |
| s-n-boc-3-bromophenylalanine, boc-l-3-bromophenylalanine, boc-3-bromo-l-phenylalanine, boc-phe 3-br-oh, boc-l-3-br-phe-oh, boc-l-3-bromophe, n-boc-3-bromo-l-phenylalanine, s-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-2-tert-butoxy carbonylamino-3-3-bromophenyl propanoic acid | |
| FBUDYESOPLBQIR-NSHDSACASA-N | |
| (2S)-3-(3-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
| 2734476 | |
| 95% |
| C14H18BrNO4 | |
| 1 g | |
| Slightly soluble in water. | |
| CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)Br)C(=O)O | |
| 344.205 | |
| 344.2 | |
| N-Boc-3-bromo-L-phenylalanine |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only