Learn More
N-BOC-3-(4-Cyanophenyl)oxaziridine, 98+%
CAS: 150884-56-3 | C13H14N2O3 | 246.27 g/mol
Supplier: Thermo Scientific Chemicals 297090020
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-BOC-3-(4-Cyanophenyl)oxaziridine | |
| 150884-56-3 | |
| 100.0 | |
| Beige to White | |
| Authentic | |
| Glass bottle | |
| MFCD00800493 | |
| n-boc-3-4-cyanophenyl oxaziridine, tert-butyl 3-4-cyanophenyl-1,2-oxaziridine-2-carboxylate, tert-butyl 3-4-cyanophenyl oxaziridine-2-carboxylate, 2-oxaziridinecarboxylicacid, 3-4-cyanophenyl-, 1,1-dimethylethyl ester, 2r,3s-rel-9ci, acmc-20n62r, n-boc-3-4-cyanophenyl oxaziridine, 98+%, 2-tert-butoxycarbonyl-3-4-cyanophenyl oxaziridine, n-tert-butoxycarbonyl-3-4-cyanophenyl-oxaziridine, 3-4-cyano-phenyl-oxaziridine-2-carboxylic acid tert-butyl ester, 3-4-cyano-phenyl oxaziridine-2-carboxylic acid tert-butyl ester | |
| ACXPNVRTMHEHMQ-UHFFFAOYSA-N | |
| tert-butyl 3-(4-cyanophenyl)oxaziridine-2-carboxylate | |
| 2725038 | |
| 98+% |
| 98+% | |
| 98.0 | |
| 57.0°C to 63.0°C | |
| 98.0% min. | |
| 98+% | |
| C13H14N2O3 | |
| 2 g | |
| Solubility in water: .. Other solubilities: soluble in ether,methylene chloride,chloroform,,etc | |
| CC(C)(C)OC(=O)N1C(O1)C2=CC=C(C=C2)C#N | |
| 246.27 | |
| 246.27 | |
| Crystalline Powder |
Chemical Identifiers
| 150884-56-3 | |
| 246.27 | |
| ACXPNVRTMHEHMQ-UHFFFAOYSA-N | |
| 2725038 | |
| CC(C)(C)OC(=O)N1C(O1)C2=CC=C(C=C2)C#N |
| C13H14N2O3 | |
| MFCD00800493 | |
| n-boc-3-4-cyanophenyl oxaziridine, tert-butyl 3-4-cyanophenyl-1,2-oxaziridine-2-carboxylate, tert-butyl 3-4-cyanophenyl oxaziridine-2-carboxylate, 2-oxaziridinecarboxylicacid, 3-4-cyanophenyl-, 1,1-dimethylethyl ester, 2r,3s-rel-9ci, acmc-20n62r, n-boc-3-4-cyanophenyl oxaziridine, 98+%, 2-tert-butoxycarbonyl-3-4-cyanophenyl oxaziridine, n-tert-butoxycarbonyl-3-4-cyanophenyl-oxaziridine, 3-4-cyano-phenyl-oxaziridine-2-carboxylic acid tert-butyl ester, 3-4-cyano-phenyl oxaziridine-2-carboxylic acid tert-butyl ester | |
| tert-butyl 3-(4-cyanophenyl)oxaziridine-2-carboxylate |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only