missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-(2-naphthyl)-L-alanine, 97%

Catalog No. AAH6320506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6320506 5 g
AAH6320503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6320506 Supplier Thermo Scientific Chemicals Supplier No. H6320506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58438-04-3 | C18H21NO4 | 315.369 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58438-04-3
Molecular Formula C18H21NO4
Molecular Weight (g/mol) 315.369
MDL Number MFCD00079671
InChI Key URKWHOVNPHQQTM-HNNXBMFYSA-N
Synonym boc-3-2-naphthyl-l-alanine, boc-2-nal-oh, boc-l-2-naphthylalanine, n-tert-butoxycarbonyl-3-2-naphthyl-l-alanine, boc-l-3-2-naphthyl-alanine, s-boc-3-2-naphthyl-l-alanine, s-n-boc-2-naphthylalanine, boc-2-l-naphthylalanine, n-boc-3-2-naphthyl-l-alanine, s-2-tert-butoxycarbonyl amino-3-naphthalen-2-yl propanoic acid
PubChem CID 7020908
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-naphthalen-2-ylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC2=CC=CC=C2C=C1)C(=O)O

Specifications

Melting Point ∼95°C
Color White
Beilstein 4326467
Quantity 5 g
Formula Weight 315.37
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Boc-3-(2-naphthyl)-L-alanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only