missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-methyl-D-serine methyl ester, 97%, Thermo Scientific Chemicals

Catalog No. p-7034723
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52421MD 250 mg
AAH5242103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52421MD Supplier Thermo Scientific Chemicals Supplier No. H52421MD

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 188476-33-7
Molecular Formula C10H19NO5
Molecular Weight (g/mol) 233.264
MDL Number MFCD06797553
InChI Key OUUNEDPIBZNRMT-SNVBAGLBSA-N
Synonym n-boc-alpha-methyl-d-serine methyl ester, n-boc-2-methyl-d-serine methyl ester, n-tert-butoxycarbonyl-2-methyl-d-serine methyl ester, r-methyl 2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoate, methyl 2r-2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoate
PubChem CID 10752097
IUPAC Name methyl (2R)-3-hydroxy-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(C)(CO)C(=O)OC

Specifications

Quantity 250 mg
Formula Weight 233.26
Percent Purity 97%
Chemical Name or Material N-Boc-2-methyl-D-serine methyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only