missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-methyl-D-serine, 97%, Thermo Scientific Chemicals

Catalog No. p-7035082
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52787MD 250 mg
AAH5278703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52787MD Supplier Thermo Scientific Chemicals Supplier No. H52787MD
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84311-18-2
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.237
MDL Number MFCD02682596
InChI Key FWRXDSRYWWYTPD-SECBINFHSA-N
Synonym r-2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoic acid, n-boc-alpha-methyl-d-serine, boc-alpha-methyl-d-ser, boc-me-d-ser-oh, 2r-2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoic acid, boc-d-alpha-methylserine, boc-a-methyl-d-ser, n-boc-a-methyl-d-serine, n-boc-2-methyl-d-serine, n-tert-butoxycarbonyl-2-methyl-d-serine
PubChem CID 24749404
IUPAC Name (2R)-3-hydroxy-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(C)(CO)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 219.24
Percent Purity 97%
Chemical Name or Material N-Boc-2-methyl-D-serine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only