missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-1-Boc-L-tryptophan, 95%
CAS: 144599-95-1 | C21H28N2O6 | 404.46 g/mol
$78.19 - $298.34
Chemical Identifiers
| CAS | 144599-95-1 |
|---|---|
| Molecular Formula | C21H28N2O6 |
| Molecular Weight (g/mol) | 404.46 |
| MDL Number | MFCD00270520 |
| InChI Key | FATGZMFSCKUQGO-HNNXBMFYSA-N |
| Synonym | boc-trp boc-oh, s-3-1-tert-butoxycarbonyl-1h-indol-3-yl-2-tert-butoxycarbonyl amino propanoic acid, boc-l-trp boc-oh, ambotzbaa1140, nalpha,1-di-boc-l-tryptophan, n,n'-di-t-butyloxycarbonyl-l-tryptophane, 2s-2-2-methylpropan-2-yl oxycarbonylamino-3-1-2-methylpropan-2-yl oxycarbonyl indol-3-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-tert-butoxycarbonyl indol-3-yl propanoic acid, 2s-3-1-tert-butoxy carbonyl-1h-indol-3-yl-2-tert-butoxy carbonyl amino propanoic acid |
| PubChem CID | 7020330 |
| IUPAC Name | (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]indol-3-yl]propanoic acid |
| SMILES | CC(C)(C)OC(=O)N[C@@H](CC1=CN(C(=O)OC(C)(C)C)C2=CC=CC=C12)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6254203
|
Thermo Scientific Chemicals
H6254203 |
1 g |
N/A
|
|
|||||
|
AAH6254206
|
Thermo Scientific Chemicals
H6254206 |
5 g |
N/A
|
|
|||||
Description
N-Boc-1-Boc-L-tryptophan is generally used as a intermediate in pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-1-Boc-L-tryptophan is generally used as a intermediate in pharmaceutical and chemical research.
Solubility
Soluble in dimethylformamide (1 mmole in 2 ml).
Notes
Store at -20°C. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
| 144599-95-1 | |
| 404.46 | |
| FATGZMFSCKUQGO-HNNXBMFYSA-N | |
| 7020330 | |
| CC(C)(C)OC(=O)N[C@@H](CC1=CN(C(=O)OC(C)(C)C)C2=CC=CC=C12)C(O)=O |
| C21H28N2O6 | |
| MFCD00270520 | |
| boc-trp boc-oh, s-3-1-tert-butoxycarbonyl-1h-indol-3-yl-2-tert-butoxycarbonyl amino propanoic acid, boc-l-trp boc-oh, ambotzbaa1140, nalpha,1-di-boc-l-tryptophan, n,n'-di-t-butyloxycarbonyl-l-tryptophane, 2s-2-2-methylpropan-2-yl oxycarbonylamino-3-1-2-methylpropan-2-yl oxycarbonyl indol-3-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-tert-butoxycarbonyl indol-3-yl propanoic acid, 2s-3-1-tert-butoxy carbonyl-1h-indol-3-yl-2-tert-butoxy carbonyl amino propanoic acid | |
| (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]indol-3-yl]propanoic acid |
Specifications
| 144599-95-1 | |
| MFCD00270520 | |
| boc-trp boc-oh, s-3-1-tert-butoxycarbonyl-1h-indol-3-yl-2-tert-butoxycarbonyl amino propanoic acid, boc-l-trp boc-oh, ambotzbaa1140, nalpha,1-di-boc-l-tryptophan, n,n'-di-t-butyloxycarbonyl-l-tryptophane, 2s-2-2-methylpropan-2-yl oxycarbonylamino-3-1-2-methylpropan-2-yl oxycarbonyl indol-3-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-tert-butoxycarbonyl indol-3-yl propanoic acid, 2s-3-1-tert-butoxy carbonyl-1h-indol-3-yl-2-tert-butoxy carbonyl amino propanoic acid | |
| FATGZMFSCKUQGO-HNNXBMFYSA-N | |
| (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]indol-3-yl]propanoic acid | |
| 7020330 | |
| 95% |
| C21H28N2O6 | |
| 1 g | |
| Soluble in dimethylformamide (1 mmole in 2 ml). | |
| CC(C)(C)OC(=O)N[C@@H](CC1=CN(C(=O)OC(C)(C)C)C2=CC=CC=C12)C(O)=O | |
| 404.46 | |
| 404.46 | |
| N-Boc-1-Boc-L-tryptophan |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only