missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1-Boc-L-tryptophan, 95%

Catalog No. AAH6254203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6254203 1 g
AAH6254206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6254203 Supplier Thermo Scientific Chemicals Supplier No. H6254203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 144599-95-1 | C21H28N2O6 | 404.46 g/mol

N-Boc-1-Boc-L-tryptophan is generally used as a intermediate in pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-1-Boc-L-tryptophan is generally used as a intermediate in pharmaceutical and chemical research.

Solubility
Soluble in dimethylformamide (1 mmole in 2 ml).

Notes
Store at -20°C. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 144599-95-1
Molecular Formula C21H28N2O6
Molecular Weight (g/mol) 404.46
MDL Number MFCD00270520
InChI Key FATGZMFSCKUQGO-HNNXBMFYSA-N
Synonym boc-trp boc-oh, s-3-1-tert-butoxycarbonyl-1h-indol-3-yl-2-tert-butoxycarbonyl amino propanoic acid, boc-l-trp boc-oh, ambotzbaa1140, nalpha,1-di-boc-l-tryptophan, n,n'-di-t-butyloxycarbonyl-l-tryptophane, 2s-2-2-methylpropan-2-yl oxycarbonylamino-3-1-2-methylpropan-2-yl oxycarbonyl indol-3-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-tert-butoxycarbonyl indol-3-yl propanoic acid, 2s-3-1-tert-butoxy carbonyl-1h-indol-3-yl-2-tert-butoxy carbonyl amino propanoic acid
PubChem CID 7020330
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]indol-3-yl]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CN(C(=O)OC(C)(C)C)C2=CC=CC=C12)C(O)=O

Specifications

Quantity 1 g
Solubility Information Soluble in dimethylformamide (1 mmole in 2 ml).
Formula Weight 404.46
Percent Purity 95%
Chemical Name or Material N-Boc-1-Boc-L-tryptophan
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only