missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-1-Aminocyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog No. p-4229094
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC440440050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC440440050 Supplier Thermo Scientific Chemicals Supplier No. 440440050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 120728-10-1 | Molecular Formula: C10H16NO4 | Molecular Weight: 214.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 120728-10-1
Molecular Formula C10H16NO4
Molecular Weight (g/mol) 214.24
MDL Number MFCD02682623
InChI Key ROVVUKFHORPDSM-UHFFFAOYSA-M
Synonym n-boc-1-aminocyclobutanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, 1-boc-amino cyclobutanecarboxylic acid, boc-cyclovaline, boc-acbc-oh, 1-n-boc-amino-cyclobutane carboxylic acid, 1-tert-butoxycarbonylamino cyclobutanecarboxylic acid, boc-1-aminocyclobutane-1-carboxylic acid, n-boc-1-aminocyclobutane carboxylic acid, boc-1-aminocyclobutanecarboxylic acid
PubChem CID 1512646
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclobutane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CCC1)C([O-])=O

Specifications

Melting Point 129°C to 133°C
CAS Min % 96.0
CAS Max % 100.0
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 5 g
Formula Weight 215.25
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-BOC-1-Aminocyclobutanecarboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF IN EYES.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you fee

RUO – Research Use Only