missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1,6-diaminohexane hydrochloride, 98%

Catalog No. AAL1948703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1948703 1 g
AAL1948706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1948703 Supplier Thermo Scientific Chemicals Supplier No. L1948703
Only null left

CAS: 65915-94-8 | C11H25ClN2O2 | 252.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65915-94-8
Molecular Formula C11H25ClN2O2
Molecular Weight (g/mol) 252.78
MDL Number MFCD00039072
InChI Key JSBWQIZQJOQPFN-UHFFFAOYSA-N
Synonym n-boc-1,6-hexanediamine hydrochloride, tert-butyl 6-aminohexyl carbamate hydrochloride, n-boc-1,6-diamino-hexane hydrochloride, tert-butyl n-6-aminohexyl carbamate hydrochloride, n-6-aminohexyl tert-butoxy carboxamide, chloride, tert-butyl 6-aminohexyl carbamate monohydrochloride, tert-butyl 6-aminohexyl carbamate hcl, acmc-1b5cm, n-1-boc-1,6-diaminohexane . hcl, n-boc-1,6-hexanediamine hcl
PubChem CID 3017638
IUPAC Name tert-butyl N-(6-aminohexyl)carbamate;hydrochloride
SMILES [H+].[Cl-].CC(C)(C)OC(=O)NCCCCCCN

Specifications

Melting Point ∽164°C (decomposition)
Beilstein 3631740
Quantity 1 g
Formula Weight 252.79
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material N-Boc-1,6-diaminohexane hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 265-976-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only