missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-1,4-butanediamine, 95%

Catalog No. AC426190050
Change view
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426190050 5 mL Glass Bottle
AC426190010 1 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC426190050 Supplier Thermo Scientific Chemicals Supplier No. 426190050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 68076-36-8 | C9H20N2O2 | 188.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68076-36-8
Molecular Formula C9H20N2O2
Molecular Weight (g/mol) 188.27
MDL Number MFCD00210019
InChI Key ZFQWJXFJJZUVPI-UHFFFAOYSA-N
Synonym tert-butyl n-4-aminobutyl carbamate, n-boc-1,4-butanediamine, n-boc-1,4-diaminobutane, tert-butyl 4-aminobutyl carbamate, tert-butyl 4-aminobutylcarbamate, boc-1,4-diaminobutane, n-boc-1,4-butandiamine, n-4-aminobutyl carbamic acid tert-butyl ester, n-tert-butoxycarbonyl-1,4-diaminobutane, mono-4-n-boc-1,4-diaminobutane
PubChem CID 4351
IUPAC Name tert-butyl N-(4-aminobutyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCN

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9840g/mL
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 5 mL
Specific Gravity 0.984
Formula Weight 188.27
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-BOC-1, 4-butanediamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only