missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-1,2-diaminoethane, 98%

Catalog No. p-3586456
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC389900010 1 g Glass Bottle
AC389900050 5 g Glass Bottle
AC389900250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC389900010 Supplier Thermo Scientific Chemicals Supplier No. 389900010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 57260-73-8 | C7H16N2O2 | 160.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57260-73-8
Molecular Formula C7H16N2O2
Molecular Weight (g/mol) 160.22
MDL Number MFCD00191871
InChI Key AOCSUUGBCMTKJH-UHFFFAOYSA-N
Synonym n-boc-ethylenediamine, tert-butyl n-2-aminoethyl carbamate, tert-butyl 2-aminoethyl carbamate, n-t-boc-ethylenediamine, n-tert-boc-ethylenediamine, tert-butyl 2-aminoethylcarbamate, n-boc-1,2-diaminoethane, n-boc-ethane-1,2-diamine, mono-n-t-boc-eda, n-boc-diaminoethane
PubChem CID 187201
IUPAC Name tert-butyl N-(2-aminoethyl)carbamate
SMILES CC(C)(C)OC(=O)NCCN

Specifications

Boiling Point 80°C to 84°C (0.3mmHg)
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.458
Linear Formula (CH3)3COC(O)NH(CH2)2NH2
Density 1.0160g/mL
Quantity 1 g
Flash Point >110°C
Formula Weight 160.22
Percent Purity 98%
Specific Gravity 1.016
Physical Form Oily Liquid
Chemical Name or Material N-BOC-1, 2-diaminoethane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only