missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonylglycine, 98+%
Description
N-Carbobenzoxyglycine is used in the dipeptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Carbobenzoxyglycine is used in the dipeptide synthesis.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Store in cool dry place. Keep away from oxidizing agents.
N-Carbobenzoxyglycine is used in the dipeptide synthesis.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Store in cool dry place. Keep away from oxidizing agents.
Specifications
Specifications
| Melting Point | 119°C to 122°C |
| Linear Formula | C6H5CH2OCONHCH2CO2H |
| Beilstein | 526877 |
| Solubility Information | Soluble in methanol. Insoluble in water. |
| Quantity | 25 g |
| Formula Weight | 209.2 |
| Percent Purity | ≥98% |
| Chemical Name or Material | N-Benzyloxycarbonylglycine |
Safety and Handling
| EINECSNumber | 214-516-0 |
| RTECSNumber | MB9129000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only