missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Benzyloxycarbonyl)sarcosine, 95%
CAS: 39608-31-6 | C11H13NO4 | 223.228 g/mol
$76.96 - $234.78
Chemical Identifiers
| CAS | 39608-31-6 |
|---|---|
| Molecular Formula | C11H13NO4 |
| Molecular Weight (g/mol) | 223.228 |
| MDL Number | MFCD00021747 |
| InChI Key | CBWFTZNMONHKNZ-UHFFFAOYSA-N |
| Synonym | z-sar-oh, cbz-sar-oh, n-benzyloxycarbonyl sarcosine, carbobenzoxy sarcosine, z-sarcosine, glycine, n-methyl-n-phenylmethoxy carbonyl, n-cbz-sarcosine, 2-benzyloxy carbonyl methyl amino acetic acid, n-carbobenzoxy sarcosine, benzyloxycarbonyl sarcosine |
| PubChem CID | 97053 |
| IUPAC Name | 2-[methyl(phenylmethoxycarbonyl)amino]acetic acid |
| SMILES | CN(CC(=O)O)C(=O)OCC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6213506
|
Thermo Scientific Chemicals
H6213506 |
5 g |
Each for $76.96
|
|
|||||
|
AAH6213514
|
Thermo Scientific Chemicals
H6213514 |
25 g |
Each for $234.78
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 39608-31-6 | |
| 223.228 | |
| CBWFTZNMONHKNZ-UHFFFAOYSA-N | |
| 97053 | |
| CN(CC(=O)O)C(=O)OCC1=CC=CC=C1 |
| C11H13NO4 | |
| MFCD00021747 | |
| z-sar-oh, cbz-sar-oh, n-benzyloxycarbonyl sarcosine, carbobenzoxy sarcosine, z-sarcosine, glycine, n-methyl-n-phenylmethoxy carbonyl, n-cbz-sarcosine, 2-benzyloxy carbonyl methyl amino acetic acid, n-carbobenzoxy sarcosine, benzyloxycarbonyl sarcosine | |
| 2-[methyl(phenylmethoxycarbonyl)amino]acetic acid |
Specifications
| 39608-31-6 | |
| MFCD00021747 | |
| CBWFTZNMONHKNZ-UHFFFAOYSA-N | |
| 2-[methyl(phenylmethoxycarbonyl)amino]acetic acid | |
| 223.228 | |
| 223.23 | |
| N-(Benzyloxycarbonyl)sarcosine |
| C11H13NO4 | |
| z-sar-oh, cbz-sar-oh, n-benzyloxycarbonyl sarcosine, carbobenzoxy sarcosine, z-sarcosine, glycine, n-methyl-n-phenylmethoxy carbonyl, n-cbz-sarcosine, 2-benzyloxy carbonyl methyl amino acetic acid, n-carbobenzoxy sarcosine, benzyloxycarbonyl sarcosine | |
| CN(CC(=O)O)C(=O)OCC1=CC=CC=C1 | |
| 5 g | |
| 97053 | |
| 95% |
Safety and Handling
TSCA : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only