missing translation for 'onlineSavingsMsg'
Learn More

N-(Benzyloxycarbonyl)sarcosine, 95%

Catalog No. AAH6213506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6213506 5 g
AAH6213514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6213506 Supplier Thermo Scientific Chemicals Supplier No. H6213506

CAS: 39608-31-6 | C11H13NO4 | 223.228 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39608-31-6
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.228
MDL Number MFCD00021747
InChI Key CBWFTZNMONHKNZ-UHFFFAOYSA-N
Synonym z-sar-oh, cbz-sar-oh, n-benzyloxycarbonyl sarcosine, carbobenzoxy sarcosine, z-sarcosine, glycine, n-methyl-n-phenylmethoxy carbonyl, n-cbz-sarcosine, 2-benzyloxy carbonyl methyl amino acetic acid, n-carbobenzoxy sarcosine, benzyloxycarbonyl sarcosine
PubChem CID 97053
IUPAC Name 2-[methyl(phenylmethoxycarbonyl)amino]acetic acid
SMILES CN(CC(=O)O)C(=O)OCC1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 223.23
Percent Purity 95%
Chemical Name or Material N-(Benzyloxycarbonyl)sarcosine
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only