missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-N-methyl-D-valine, 95%, Thermo Scientific™

Catalog No. p-7056550
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6553106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6553106 Supplier Thermo Scientific Chemicals Supplier No. H6553106

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 53978-73-7
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD04973293
InChI Key NNEHOKZDWLJKHP-GFCCVEGCSA-N
Synonym z-d-n-me-val-oh, n-cbz-n-methyl-d-valine, r-2-benzyloxy carbonyl methyl amino-3-methylbutanoic acid, ambotzzaa1004, benzyloxycarbonyl-n-alpha-methyl-d-valine, n-benzyloxy carbonyl-n-methyl-d-valine, d-valine, n-methyl-n-phenylmethoxy carbonyl, 2r-2-benzyloxy carbonyl methyl amino-3-methylbutanoic acid
PubChem CID 7018026
IUPAC Name (2R)-2-{[(benzyloxy)carbonyl](methyl)amino}-3-methylbutanoic acid
SMILES CC(C)[C@@H](N(C)C(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Color White
Beilstein 3593385
Quantity 5 g
Formula Weight 265.31
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Benzyloxycarbonyl-N-methyl-D-valine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold