missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-proline, 98+%

CAS: 1148-11-4 | C13H15NO4 | 249.266 g/mol

Supplier:  Thermo Scientific Chemicals B2167218

 View more versions of this product

Catalog No. AAB2167218


Only null left
Explore available promotions

Description

Description

N-(Benzyloxycarbonyl)-L-proline is a potent inhibitor of prolidase; a specific peptidase that cleaves dipeptides with a C-terminal prolyl and hydroxylprolyl residue. It is also used in the synthesis of N-(L-Prolyl)-β-alanine which is a derivative of the naturally occurring beta amino acid β-Alanine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(Benzyloxycarbonyl)-L-proline is a potent inhibitor of prolidase; a specific peptidase that cleaves dipeptides with a C-terminal prolyl and hydroxylprolyl residue. It is also used in the synthesis of N-(L-Prolyl)-β-alanine which is a derivative of the naturally occurring beta amino acid β-Alanine.

Solubility
Solubility in methanol, almost transparency.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

N-Benzyloxycarbonyl-L-proline
75°C to 77°C
MFCD00003170
88579
JXGVXCZADZNAMJ-NSHDSACASA-N
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O
249.266
249.27
1148-11-4
C13H15NO4
50 g
z-pro-oh, n-benzyloxycarbonyl-l-proline, n-carbobenzoxy-l-proline, n-cbz-l-proline, benzyloxycarbonyl-l-proline, carbobenzoxyproline, carbobenzyloxy-l-proline, carbobenzoxy-l-proline, benzyloxycarbonylproline, n-carbobenzyloxy-l-proline
−60° (c=1 in Acetic Acid)
(2S)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
101987
≥98%

Chemical Identifiers

1148-11-4
249.266
JXGVXCZADZNAMJ-NSHDSACASA-N
101987
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O
C13H15NO4
MFCD00003170
z-pro-oh, n-benzyloxycarbonyl-l-proline, n-carbobenzoxy-l-proline, n-cbz-l-proline, benzyloxycarbonyl-l-proline, carbobenzoxyproline, carbobenzyloxy-l-proline, carbobenzoxy-l-proline, benzyloxycarbonylproline, n-carbobenzyloxy-l-proline
(2S)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-557-4

RTECSNumber : UY0745000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only