missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-phenylalanine, 99%

Catalog No. p-7023532
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1718206 5 g
AAA1718214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1718206 Supplier Thermo Scientific Chemicals Supplier No. A1718206

CAS: 1161-13-3 | C17H17NO4 | 299.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1161-13-3
Molecular Formula C17H17NO4
Molecular Weight (g/mol) 299.33
MDL Number MFCD00020418
InChI Key RRONHWAVOYADJL-HNNXBMFYSA-N
Synonym z-phe-oh, n-cbz-l-phenylalanine, carbobenzoxyphenylalanine, cbz-phe-oh, n-carbobenzoxy-l-phenylalanine, carbobenzoxy-l-phenylalanine, z-l-phenylalanine, benzyloxycarbonyl phenylalanine, n-carbobenzyloxy-l-phenylalanine
PubChem CID 70878
IUPAC Name (2S)-3-phenyl-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES OC(=O)[C@H](CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 86°C to 92°C
Quantity 5 g
Beilstein 2222826
Optical Rotation +5° (c=5 in Acetic Acid)
Formula Weight 299.33
Percent Purity 99%
Chemical Name or Material N-Benzyloxycarbonyl-L-phenylalanine
EINECSNumber 214-599-3
RTECSNumber AY4343000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only