Learn More
N-Benzyloxycarbonyl-L-glutamic acid 5-tert-butyl ester, 97%, Thermo Scientific Chemicals
$110.33 - $110.33
Chemical Identifiers
| CAS | 3886-08-6 |
|---|---|
| Molecular Formula | C17H22NO6 |
| Molecular Weight (g/mol) | 336.37 |
| MDL Number | MFCD00038274 |
| InChI Key | GLMODRZPPBZPPB-ZDUSSCGKSA-M |
| Synonym | z-glu otbu-oh, cbz-glu obut-oh, cbz-l-glu otbu-oh, s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, n-cbz-l-glutamic acid 5-tert-butyl ester, n-benzyloxycarbonyl-l-glutamic acid 5-tert-butyl ester, 2s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, 2s-4-tert-butyl oxycarbonyl-2-phenylmethoxy carbonylamino butanoic acid, 5-tert-butyl n-carbobenzoxy-l-glutamate, pubchem6261 |
| PubChem CID | 107472 |
| IUPAC Name | (2S)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid |
| SMILES | CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2255706
|
Thermo Scientific Chemicals
B2255706 |
5 g |
Each for $110.33
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3886-08-6 | |
| 336.37 | |
| GLMODRZPPBZPPB-ZDUSSCGKSA-M | |
| 107472 | |
| CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1 |
| C17H22NO6 | |
| MFCD00038274 | |
| z-glu otbu-oh, cbz-glu obut-oh, cbz-l-glu otbu-oh, s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, n-cbz-l-glutamic acid 5-tert-butyl ester, n-benzyloxycarbonyl-l-glutamic acid 5-tert-butyl ester, 2s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, 2s-4-tert-butyl oxycarbonyl-2-phenylmethoxy carbonylamino butanoic acid, 5-tert-butyl n-carbobenzoxy-l-glutamate, pubchem6261 | |
| (2S)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid |
Specifications
| 3886-08-6 | |
| C17H22NO6 | |
| 5 g | |
| GLMODRZPPBZPPB-ZDUSSCGKSA-M | |
| CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1 | |
| 336.37 | |
| 337.37 | |
| N-Benzyloxycarbonyl-L-glutamic acid 5-tert-butyl ester |
| 85°C to 88°C | |
| MFCD00038274 | |
| z-glu otbu-oh, cbz-glu obut-oh, cbz-l-glu otbu-oh, s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, n-cbz-l-glutamic acid 5-tert-butyl ester, n-benzyloxycarbonyl-l-glutamic acid 5-tert-butyl ester, 2s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, 2s-4-tert-butyl oxycarbonyl-2-phenylmethoxy carbonylamino butanoic acid, 5-tert-butyl n-carbobenzoxy-l-glutamate, pubchem6261 | |
| −13° (c=2 in Methanol) | |
| (2S)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid | |
| 107472 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 223-421-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only