missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-glutamic acid 1-methyl ester, 98%

Catalog No. AAH5341703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5341703 1 g
AAH5341706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5341703 Supplier Thermo Scientific Chemicals Supplier No. H5341703

CAS: 5672-83-3 | C14H17NO6 | 295.291 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5672-83-3
Molecular Formula C14H17NO6
Molecular Weight (g/mol) 295.291
MDL Number MFCD00083278
InChI Key BGMCTGARFXPQML-NSHDSACASA-N
Synonym z-glu-ome, z-l-glu-ome, 1-methyl n-carbobenzoxy-l-glutamate, 4s-4-benzyloxy carbonyl amino-5-methoxy-5-oxopentanoic acid, cbz-glu-ome, l-glutamic acid,n-phenylmethoxy carbonyl-, 1-methyl ester, n-cbz-l-glu-ome, z-l-glutamic acid 1-methyl ester, n-cbz-l-glutamic acid 1-methyl ester, n-carbobenzoxy-l-glutamic acid 1-methyl ester
PubChem CID 2733408
IUPAC Name (4S)-5-methoxy-5-oxo-4-(phenylmethoxycarbonylamino)pentanoic acid
SMILES COC(=O)C(CCC(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 64°C to 70°C
Refractive Index 1.4365
Quantity 1 g
Beilstein 2567599
Optical Rotation −24° (c=1 in Ethanol)
Formula Weight 295.29
Percent Purity 98%
Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid 1-methyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only