missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester, 95%

Catalog No. p-7041655
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6297506 5 g
AAH6297514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6297506 Supplier Thermo Scientific Chemicals Supplier No. H6297506

CAS: 3705-42-8 | C20H21NO6 | 371.389 g/mol

N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester used as a potential neuroprotective agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester used as a potential neuroprotective agent.

Solubility
Soluble in methanol, dimethyl sulfoxide. Slightly soluble in water.

Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 3705-42-8
Molecular Formula C20H21NO6
Molecular Weight (g/mol) 371.389
MDL Number MFCD00077013
InChI Key VWHKODOUMSMUAF-KRWDZBQOSA-N
Synonym z-glu-obzl, cbz-glu-obzl, cbz-l-glutamic acid 1-benzyl ester, z-l-glu-obzl, n-cbz-l-glutamic acid benzyl ester, n-carbobenzyloxy-l-glutamic acid o-benzyl ester, s-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, 4s-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, n-benzyloxycarbonyl-l-glutamic acid, a-benzyl ester, z-l-glutamic acid alpha-benzyl ester
PubChem CID 6994031
IUPAC Name (4S)-5-oxo-5-phenylmethoxy-4-(phenylmethoxycarbonylamino)pentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 98°C to 102°C
Quantity 5 g
Solubility Information Soluble in methanol,dimethyl sulfoxide. Slightly soluble in water.
Formula Weight 371.38
Percent Purity 95%
Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only