missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-aspartic acid 4-benzyl ester, 98+%, Thermo Scientific™

$206.53 - $206.53

Chemical Identifiers

CAS 3479-47-8
Molecular Formula C19H19NO6
Molecular Weight (g/mol) 357.36
MDL Number MFCD00037820
InChI Key VUKCNAATVIWRTF-UHFFFAOYNA-N
Synonym z-asp obzl-oh, cbz-asp obzl-oh, n-cbz-l-aspartic acid 4-benzyl ester, z-l-aspartic acid 4-benzyl ester, n-benzyloxycarbonyl-l-aspartic acid 4-benzyl ester, 4-benzyl hydrogen n-phenylmethoxy carbonyl-l-aspartate, s-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid, n-carbobenzoxy-l-aspartic acid 4-benzyl ester, 2s-4-oxo-4-phenylmethoxy-2-phenylmethoxycarbonylamino butanoic acid, 2s-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid
PubChem CID 47602
IUPAC Name (2S)-4-oxo-4-phenylmethoxy-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0843606
View Documents
Thermo Scientific Chemicals
L0843606
5 g
Each for $206.53
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is widely used as a pharmaceutical intermediate and in food industry.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

3479-47-8
357.36
VUKCNAATVIWRTF-UHFFFAOYNA-N
47602
OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
C19H19NO6
MFCD00037820
z-asp obzl-oh, cbz-asp obzl-oh, n-cbz-l-aspartic acid 4-benzyl ester, z-l-aspartic acid 4-benzyl ester, n-benzyloxycarbonyl-l-aspartic acid 4-benzyl ester, 4-benzyl hydrogen n-phenylmethoxy carbonyl-l-aspartate, s-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid, n-carbobenzoxy-l-aspartic acid 4-benzyl ester, 2s-4-oxo-4-phenylmethoxy-2-phenylmethoxycarbonylamino butanoic acid, 2s-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid
(2S)-4-oxo-4-phenylmethoxy-2-(phenylmethoxycarbonylamino)butanoic acid

Specifications

3479-47-8
C19H19NO6
5 g
z-asp obzl-oh, cbz-asp obzl-oh, n-cbz-l-aspartic acid 4-benzyl ester, z-l-aspartic acid 4-benzyl ester, n-benzyloxycarbonyl-l-aspartic acid 4-benzyl ester, 4-benzyl hydrogen n-phenylmethoxy carbonyl-l-aspartate, s-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid, n-carbobenzoxy-l-aspartic acid 4-benzyl ester, 2s-4-oxo-4-phenylmethoxy-2-phenylmethoxycarbonylamino butanoic acid, 2s-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid
VUKCNAATVIWRTF-UHFFFAOYNA-N
(2S)-4-oxo-4-phenylmethoxy-2-(phenylmethoxycarbonylamino)butanoic acid
47602
≥98%
105°C to 107°C
MFCD00037820
2065292
Insoluble in water.
OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
357.36
357.37
N-Benzyloxycarbonyl-L-aspartic acid 4-benzyl ester
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 222-456-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'