missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-alaninol

Catalog No. AAH2706603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2706603 1 g
AAH2706606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2706603 Supplier Thermo Scientific Chemicals Supplier No. H2706603

CAS: 66674-16-6 | C11H15NO3 | 209.25 g/mol

N-Benzyloxycarbonyl-L-alaninol is used as an important organic intermediate. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzyloxycarbonyl-L-alaninol is used as an important organic intermediate. It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Chemical Identifiers

CAS 66674-16-6
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.25
MDL Number MFCD00672530
InChI Key AFPHMHSLDRPUSM-VIFPVBQESA-N
Synonym cbz-l-alaninol, s-benzyl 1-hydroxypropan-2-yl carbamate, n-z-l-alaninol, s-benzyl 1-hydroxypropan-2-ylcarbamate, benzyl n-2s-1-hydroxypropan-2-yl carbamate, carbamic acid, 1s-2-hydroxy-1-methylethyl-, phenylmethyl ester, phenylmethyl n-2s-1-oxidanylpropan-2-yl carbamate, n-2s-1-hydroxypropan-2-yl carbamic acid phenylmethyl ester, n-cbz-l-alaninol, cbz-l-ala-ol
PubChem CID 10262499
IUPAC Name benzyl N-[(2S)-1-hydroxypropan-2-yl]carbamate
SMILES C[C@@H](CO)NC(=O)OCC1=CC=CC=C1

Specifications

Solubility Information Soluble in ethanol.
Quantity 1 g
Formula Weight 209.25
Chemical Name or Material N-Benzyloxycarbonyl-L-alaninol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only