missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-alanine, 98%

Catalog No. aaa1671606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1671606 5 g
AAA1671614 25 g
AAA1671622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1671606 Supplier Thermo Scientific Chemicals Supplier No. A1671606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1142-20-7 | C11H13NO4 | 223.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1142-20-7
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00002640
InChI Key TYRGLVWXHJRKMT-QMMMGPOBSA-N
Synonym n-carbobenzyloxy-l-alanine, z-ala-oh, n-cbz-l-alanine, z-l-alanine, n-benzyloxycarbonyl-l-alanine, cbz-l-alanine, cbz-ala-oh, carbobenzyloxy-l-alanine, benzyloxycarbonyl-l-alanine, carbobenzoxy-l-alanine
PubChem CID 736104
SMILES C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 85°C to 87°C
Quantity 5 g
Beilstein 2056164
Optical Rotation −14.5° (c=2 in Acetic Acid)
IUPAC Name (2S)-2-(phenylmethoxycarbonylamino)propanoic acid
Formula Weight 223.23
Percent Purity 98%
Chemical Name or Material N-Benzyloxycarbonyl-L-alanine
EINECSNumber 214-532-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only