missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-D-glutamic acid 1-benzyl ester, 97%

Catalog No. AAH5512106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5512106 5 g
AAH5512103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5512106 Supplier Thermo Scientific Chemicals Supplier No. H5512106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 65706-99-2 | C20H21NO6 | 371.389 g/mol

N-Benzyloxycarbonyl-D-glutamic acid 1-benzyl ester, is used as a reagent in the synthesis of acyl peptides that act as immunostimulants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzyloxycarbonyl-D-glutamic acid 1-benzyl ester, is used as a reagent in the synthesis of acyl peptides that act as immunostimulants.

Solubility
Soluble in methanol and acetic acid.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 65706-99-2
Molecular Formula C20H21NO6
Molecular Weight (g/mol) 371.389
MDL Number MFCD00069647
InChI Key VWHKODOUMSMUAF-QGZVFWFLSA-N
Synonym z-d-glu-obzl, cbz-d-glu-obzl, n-cbz-d-glutamic acid alpha-benzyl ester, r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, 1-benzyl n-carbobenzoxy-d-glutamate, 4r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, z-d-glu-obn, cbz-d-glu-och2ph, pubchem14966, cbz-d-glu oh-obzl
PubChem CID 7018162
IUPAC Name (4R)-5-oxo-5-phenylmethoxy-4-(phenylmethoxycarbonylamino)pentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2=CC=CC=C2

Specifications

Quantity 5 g
Beilstein 5305848
Solubility Information Soluble in methanol and acetic acid.
Formula Weight 371.39
Percent Purity 97%
Chemical Name or Material N-Benzyloxycarbonyl-D-glutamic acid 1-benzyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only