missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-D-alaninol, 98%

Catalog No. AAB2237214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2237214 25 g
AAB2237203 1 g
AAB2237206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2237214 Supplier Thermo Scientific Chemicals Supplier No. B2237214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 61425-27-2 | C11H15NO3 | 209.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61425-27-2
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.25
MDL Number MFCD00672531
InChI Key AFPHMHSLDRPUSM-SECBINFHSA-N
Synonym r-benzyl 1-hydroxypropan-2-yl carbamate, cbz-d-alaninol, z-d-ala-ol, n-z-d-alaninol, r-2-cbz-amino-1-propanol, n-benzyloxycarbonyl-d-alaninol, benzyl n-2r-1-hydroxypropan-2-yl carbamate, benzyl 1r-2-hydroxy-1-methylethylcarbamate, n-1r-2-hydroxy-isopropyl phenylmethoxy carboxamide
PubChem CID 6951254
IUPAC Name benzyl N-[(2R)-1-hydroxypropan-2-yl]carbamate
SMILES C[C@H](CO)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 82°C to 84°C
Quantity 25 g
Formula Weight 209.25
Percent Purity 98%
Chemical Name or Material N-Benzyloxycarbonyl-D-alaninol
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only