missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylidenemethylamine, 99%

Catalog No. AC106010050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC106010050 5 g
AC106010250 25 g
AC106011000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC106010050 Supplier Thermo Scientific Chemicals Supplier No. 106010050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 622-29-7 | C8H9N | 119.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 622-29-7
MDL Number MFCD00008294
InChI Key HXTGGPKOEKKUQO-UHFFFAOYSA-N
Synonym n-benzylidenemethylamine, n-benzylidenemethanamine, benzylidenemethylamine, n-methylbenzaldimine, methanamine, n-phenylmethylene, benzylidenemethanamine, n-methylbenzylideneamine, methylamine, n-benzylidene, n-phenylmethylene methanamine, benzylidene-methyl-amine
PubChem CID 73954
IUPAC Name N-methyl-1-phenylmethanimine
SMILES CN=CC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Density 96%g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H9N
Refractive Index 1.5500 to 1.5540 (20°C, 589nm)
Linear Formula C6H5CH=NCH3
Quantity 5 g
Beilstein 07, 213
Molecular Weight (g/mol) 119.167
Formula Weight 119.17
Percent Purity ≥98.5% (GC)
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material N-Benzylidenemethylamine, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-728-2
TSCA TSCA

RUO – Research Use Only