missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylidenemethylamine, 98+%

Catalog No. p-7028045
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
AAB2516106 5 g
AAB2516114 25 g
AAB2516122 100 g
3 options

Catalog No. AAB2516106

Supplier: Thermo Scientific Chemicals B2516106

Only null left

CAS: 622-29-7 | C8H9N | 119.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 622-29-7
Molecular Formula C8H9N
Molecular Weight (g/mol) 119.167
MDL Number MFCD00008294
InChI Key HXTGGPKOEKKUQO-UHFFFAOYSA-N
Synonym n-benzylidenemethylamine, n-benzylidenemethanamine, benzylidenemethylamine, n-methylbenzaldimine, methanamine, n-phenylmethylene, benzylidenemethanamine, n-methylbenzylideneamine, methylamine, n-benzylidene, n-phenylmethylene methanamine, benzylidene-methyl-amine
PubChem CID 73954
IUPAC Name N-methyl-1-phenylmethanimine
SMILES CN=CC1=CC=CC=C1

Specifications

Density 0.967
Boiling Point 66°C to 69°C (10 mmHg)
Flash Point 77°C (170°F)
Refractive Index 1.552
Quantity 5 g
Beilstein 878278
Formula Weight 119.17
Percent Purity ≥98%
Chemical Name or Material N-Benzylidenemethylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-728-2
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only