missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylidenemethylamine, 98+%

Catalog No. AAB2516106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2516106 5 g
AAB2516114 25 g
AAB2516122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2516106 Supplier Thermo Scientific Chemicals Supplier No. B2516106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 622-29-7 | C8H9N | 119.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 622-29-7
Molecular Formula C8H9N
Molecular Weight (g/mol) 119.167
MDL Number MFCD00008294
InChI Key HXTGGPKOEKKUQO-UHFFFAOYSA-N
Synonym n-benzylidenemethylamine, n-benzylidenemethanamine, benzylidenemethylamine, n-methylbenzaldimine, methanamine, n-phenylmethylene, benzylidenemethanamine, n-methylbenzylideneamine, methylamine, n-benzylidene, n-phenylmethylene methanamine, benzylidene-methyl-amine
PubChem CID 73954
IUPAC Name N-methyl-1-phenylmethanimine
SMILES CN=CC1=CC=CC=C1

Specifications

Density 0.967
Boiling Point 66°C to 69°C (10 mmHg)
Flash Point 77°C (170°F)
Refractive Index 1.552
Quantity 5 g
Beilstein 878278
Formula Weight 119.17
Percent Purity ≥98%
Chemical Name or Material N-Benzylidenemethylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-728-2
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only