missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylhydroxylamine Hydrochloride 98.0+%, TCI America™

Catalog No. B40735G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40735G 5 g
B407325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40735G Supplier TCI America Supplier No. B40735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29601-98-7
Molecular Formula C7H10ClNO
Molecular Weight (g/mol) 159.613
MDL Number MFCD00043377
InChI Key YSNXOQGDHGUKCZ-UHFFFAOYSA-N
Synonym n-benzylhydroxylamine hydrochloride, n-benzylhydroxylamine hcl, n-benzyl-n-hydroxylamine hydrochloride, benzenemethanamine, n-hydroxy-, hydrochloride, n-benzylhydroxyamine hydrochloride, bnnhoh*hydrochloride;, acmc-1cr2p, ksc493k8t, benzylhydroxyamine hydrochloride, n-benzylhydroxylamne hydrochlorde
PubChem CID 11332622
IUPAC Name N-benzylhydroxylamine;hydrochloride
SMILES C1=CC=C(C=C1)CNO.Cl

Specifications

Melting Point 106°C
Color White
Quantity 5 g
Formula Weight 159.61
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzylhydroxylamine Hydrochloride