missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylhydroxylamine hydrochloride, 98%

Catalog No. AC391040050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC391040050 5 g
AC391040250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC391040050 Supplier Thermo Scientific Chemicals Supplier No. 391040050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 29601-98-7 | C7H9NO·ClH | 159.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29601-98-7
Molecular Formula C7H9NO·ClH
Molecular Weight (g/mol) 159.62
InChI Key YSNXOQGDHGUKCZ-UHFFFAOYSA-N
Synonym n-benzylhydroxylamine hydrochloride, n-benzylhydroxylamine hcl, n-benzyl-n-hydroxylamine hydrochloride, benzenemethanamine, n-hydroxy-, hydrochloride, n-benzylhydroxyamine hydrochloride, bnnhoh*hydrochloride;, acmc-1cr2p, ksc493k8t, benzylhydroxyamine hydrochloride, n-benzylhydroxylamne hydrochlorde
PubChem CID 11332622
IUPAC Name N-benzylhydroxylamine;hydrochloride
SMILES C1=CC=C(C=C1)CNO.Cl

Specifications

Melting Point 108.0°C to 110.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (Argentometry)
Linear Formula C6H5CH2NHOH·HCl
Quantity 5 g
Formula Weight 159.62
Percent Purity 98%
Chemical Name or Material N-Benzylhydroxylamine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only