missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylcinchonidinium Chloride 98.0+%, TCI America™

Catalog No. b168310g
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B168310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B168310G Supplier TCI America Supplier No. B168310G
Only null left

Chemical Identifiers

CAS 69257-04-1
Molecular Formula C26H29ClN2O
Molecular Weight (g/mol) 420.981
MDL Number MFCD00082422
InChI Key FCHYSBWCOKEPNQ-UHOREHQSSA-M
Synonym BCDC
PubChem CID 71306949
IUPAC Name (S)-[(2R,4R,5S)-1-benzyl-5-ethenyl-1-azoniabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethanol;chloride
SMILES C=CC1C[N+]2(CCC1CC2C(C3=CC=NC4=CC=CC=C34)O)CC5=CC=CC=C5.[Cl-]

Specifications

Quantity 10 g
Formula Weight 420.98
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzylcinchonidinium Chloride [Chiral Phase-Transfer Catalyst]